Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029088.2 | NP_001024259.1 | 12 - 13 | 2182 | 75 | |
NM_001269873.3 | NP_001256802.1 | 13 - 14 | 2203 | 75 | ||
NM_001269875.4 | NP_001256804.1 | 13 - 14 | 2227 | 75 | ||
NM_001269876.3 | NP_001256805.1 | 12 - 13 | 2143 | 75 | ||
NM_001269878.3 | NP_001256807.1 | 13 - 14 | 2203 | 75 | ||
NM_001269880.3 | NP_001256809.1 | 13 - 14 | 2182 | 75 | ||
GenBank mRNA | AF431891.1 | - | 12 - 13 | 2143 | 75 | |
JQ582463.1 | - | 13 - 14 | 2203 | 75 | ||
JQ606809.1 | - | 13 - 14 | 2182 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |