Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029536.5 | NP_001024707.1 | 4 - 5 | 1760 | 73 | |
NM_001381134.1 | NP_001366990.1 | 4 - 5 | 1760 | 73 | ||
GenBank mRNA | AF200706.1 | - | 4 - 5 | 1767 | 73 | |
AF200707.1 | - | 4 - 5 | 1765 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |