Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpdh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129238.4 | NP_001122710.1 | 2 - 3 | 219 | 141 | |
NM_066787.6 | NP_499188.2 | 3 - 4 | 328 | 141 | ||
NM_181903.7 | NP_871632.1 | 3 - 4 | 385 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |