Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpdh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129238.4 | NP_001122710.1 | 4 - 5 | 385 | 115 | |
NM_181903.7 | NP_871632.1 | 5 - 6 | 551 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |