Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zak-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372611.2 | NP_001360014.1 | 6 - 7 | 1125 | 77 | |
NM_001379778.1 | NP_001367790.1 | 6 - 7 | 1143 | 77 | ||
NM_001379779.2 | NP_001367792.1 | 6 - 7 | 1148 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |