Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065059.9 | NP_497460.2 | 2 - 3 | 185 | 72 | |
NM_181914.6 | NP_871643.1 | 2 - 3 | 187 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |