Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xbp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083177.4 | NP_001076646.1 | 1 - 2 | 427 | 64 | |
GenBank mRNA | AF443190.1 | - | 1 - 2 | 286 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |