Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtm-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027431.6 | NP_001022602.1 | 2 - 3 | 255 | 95 | |
NM_067352.6 | NP_499753.1 | 2 - 3 | 255 | 95 | ||
NM_181951.6 | NP_871680.1 | 2 - 3 | 570 | 95 | ||
GenBank mRNA | AY313177.1 | - | 2 - 3 | 235 | 95 | |
AY313179.1 | - | 2 - 3 | 553 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |