Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065706.5 | NP_498107.2 | 3 - 4 | 302 | 96 | |
NM_181964.4 | NP_871693.1 | 3 - 4 | 324 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |