Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ooc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064055.4 | NP_496456.1 | 5 - 6 | 972 | 144 | |
NM_182110.3 | NP_871910.1 | 5 - 6 | 959 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |