Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373792.1 | NP_001359656.1 | 1 - 2 | 127 | 59 | |
NM_001381607.1 | NP_001367045.1 | 1 - 2 | 133 | 59 | ||
NM_064041.5 | NP_496442.1 | 1 - 2 | 127 | 59 | ||
NR_138514.1 | 1 - 2 | 745 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |