Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027178.4 | NP_001022349.1 | 2 - 3 | 178 | 82 | |
NM_063499.7 | NP_495900.1 | 2 - 3 | 199 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |