Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027178.4 | NP_001022349.1 | 3 - 4 | 1125 | 68 | |
NM_063499.7 | NP_495900.1 | 3 - 4 | 1146 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |