Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383861.1 | 16 - 17 | 3048 | 123 | ||
NM_062856.7 | NP_495257.3 | 16 - 17 | 3088 | 123 | ||
NM_182120.5 | NP_871920.1 | 5 - 6 | 1226 | 123 | ||
GenBank mRNA | AY601656.1 | - | 5 - 6 | 1205 | 123 | |
AY601657.1 | - | 5 - 6 | 1134 | 123 | ||
AY601658.1 | - | 4 - 5 | 1053 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |