Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063871.6 | NP_496272.2 | 2 - 3 | 816 | 65 | |
NM_182124.7 | NP_871924.2 | 3 - 4 | 766 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |