Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267523.2 | NP_001254452.1 | 2 - 3 | 463 | 79 | |
NM_001306640.4 | NP_001293569.1 | 3 - 4 | 458 | 79 | ||
NM_001306641.2 | NP_001293570.1 | 2 - 3 | 463 | 79 | ||
NM_001306642.3 | NP_001293571.1 | 5 - 6 | 699 | 79 | ||
NM_001306643.3 | NP_001293572.1 | 4 - 5 | 540 | 79 | ||
NM_001381668.1 | NP_001367248.1 | 5 - 6 | 699 | 79 | ||
NM_001381670.1 | NP_001368261.1 | 3 - 4 | 399 | 79 | ||
NM_001383996.1 | 5 - 6 | 763 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |