Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047333.6 | NP_001040798.1 | 3 - 4 | 443 | 79 | |
NM_001306543.3 | NP_001293472.1 | 3 - 4 | 296 | 79 | ||
NM_001383892.1 | 3 - 4 | 356 | 79 | |||
NM_063199.6 | NP_495600.1 | 3 - 4 | 378 | 79 | ||
NM_182144.6 | NP_871944.1 | 4 - 5 | 1056 | 79 | ||
NM_182145.5 | NP_871945.1 | 1 - 2 | 549 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |