Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381541.1 | NP_001367041.1 | 5 - 6 | 1777 | 73 | |
NM_001381542.2 | NP_001366731.1 | 5 - 6 | 1783 | 73 | ||
NM_001381543.2 | NP_001367082.1 | 4 - 5 | 1677 | 73 | ||
GenBank mRNA | AY314770.1 | - | 5 - 6 | 1783 | 73 | |
AY314771.1 | - | 5 - 6 | 1777 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |