Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383830.1 | 2 - 3 | 223 | 87 | ||
NM_062589.9 | NP_494990.2 | 2 - 3 | 235 | 87 | ||
NM_062590.4 | NP_494991.1 | 2 - 3 | 276 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |