Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129133.4 | NP_001122605.1 | 2 - 3 | 182 | 61 | |
NM_182199.7 | NP_871999.2 | 2 - 3 | 182 | 61 |
Gene Symbol: F14D2.15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026913.3 | NP_001022084.1 | 2 - 3 | 183 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |