Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381336.1 | NP_001367464.1 | 2 - 3 | 827 | 117 | |
NM_001383792.1 | 3 - 4 | 595 | 117 |
Gene Symbol: F36H5.14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383791.1 | 3 - 4 | 628 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |