Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctns-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063303.6 | NP_495704.1 | 2 - 3 | 334 | 98 | |
NM_182222.6 | NP_872022.1 | 2 - 3 | 336 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |