Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ce02422393_g1
See other WNK-1 GE Assays ›
Gene Symbol:
wnk-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C46C2.1
Chromosome Location:
Chr.IV: 9200102 - 9212242 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID
Ce02422393_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: wnk-1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001268439.4
NP_001255368.1
9 - 10
3576
73
NM_001268440.3
NP_001255369.1
8 - 9
3255
73
NM_001268441.3
NP_001255370.1
9 - 10
3584
73
NM_001268442.4
NP_001255371.1
8 - 9
3255
73
NM_001268443.3
NP_001255372.1
8 - 9
3249
73
NM_001268444.3
NP_001255373.1
8 - 9
3249
73
NM_001268445.3
NP_001255374.1
6 - 7
3096
73
NM_001268446.3
NP_001255375.1
6 - 7
3096
73
NM_001268447.3
NP_001255376.1
6 - 7
3090
73
NM_001268448.3
NP_001255377.1
6 - 7
3090
73
NM_001307103.3
NP_001294032.1
9 - 10
3580
73
NM_001307104.3
NP_001294033.1
8 - 9
3255
73
NM_001307106.4
NP_001294035.1
8 - 9
3255
73
NM_001307107.3
NP_001294036.1
9 - 10
3570
73
NM_001307108.3
NP_001294037.1
8 - 9
3249
73
NM_001307109.4
NP_001294038.1
9 - 10
3566
73
NM_001380399.1
NP_001368454.1
9 - 10
3576
73
NM_001380400.1
NP_001368092.1
9 - 10
3570
73
NM_069202.6
NP_501603.3
9 - 10
3564
73
NM_182275.7
NP_872075.2
9 - 10
3566
73
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Detect Genomic DNA
Caenorhabditis elegans
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein phosphorylation
hyperosmotic response
multicellular organism development
negative regulation of sodium ion transport
phosphorylation
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
ion homeostasis
multicellular organismal water homeostasis
positive regulation of potassium ion import
positive regulation of sodium ion transmembrane transporter activity
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
chloride channel inhibitor activity
potassium channel inhibitor activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0