Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268365.3 | NP_001255294.1 | 28 - 29 | 4782 | 71 | |
NM_001268366.3 | NP_001255295.1 | 23 - 24 | 3988 | 71 | ||
NM_001268369.2 | NP_001255298.1 | 24 - 25 | 3976 | 71 | ||
NM_001307278.3 | NP_001294207.1 | 24 - 25 | 4186 | 71 | ||
NM_001372869.1 | NP_001359805.1 | 24 - 25 | 3976 | 71 | ||
NM_001380324.1 | NP_001367366.1 | 25 - 26 | 4168 | 71 | ||
NM_001380325.1 | NP_001367367.1 | 27 - 28 | 4409 | 71 | ||
NM_001380326.1 | NP_001367365.1 | 5 - 6 | 770 | 71 | ||
NM_068785.7 | NP_501186.5 | 24 - 25 | 4135 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |