Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182313.1 | NP_872113.1 | 1 - 2 | 163 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |