Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276947.3 | NP_001263876.1 | 12 - 13 | 1564 | 150 | |
NM_001276948.3 | NP_001263877.1 | 12 - 13 | 1537 | 150 | ||
NM_001276949.3 | NP_001263878.1 | 9 - 10 | 1306 | 150 | ||
NM_182337.7 | NP_872137.2 | 13 - 14 | 1644 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |