Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026422.6 | NP_001021593.1 | 1 - 2 | 638 | 142 | |
GenBank mRNA | AF020344.1 | - | 1 - 2 | 687 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |