Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026425.6 | NP_001021596.1 | 5 - 6 | 511 | 81 | |
NM_001026427.7 | NP_001021598.2 | 6 - 7 | 601 | 81 | ||
NM_001377627.2 | NP_001364785.1 | 4 - 5 | 580 | 81 | ||
NM_001377628.3 | NP_001364784.1 | 4 - 5 | 574 | 81 | ||
NM_001381205.1 | NP_001367893.1 | 6 - 7 | 673 | 81 | ||
NM_001381206.1 | NP_001368498.1 | 6 - 7 | 595 | 81 | ||
NM_001381207.1 | NP_001368499.1 | 5 - 6 | 505 | 81 | ||
NM_001381208.1 | NP_001368501.1 | 3 - 4 | 271 | 81 | ||
NM_001381209.1 | NP_001368502.1 | 3 - 4 | 265 | 81 | ||
GenBank mRNA | AF020342.1 | - | 4 - 5 | 878 | 81 | |
AF020343.1 | - | 4 - 5 | 870 | 81 | ||
AF032112.1 | - | 4 - 5 | 913 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |