Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377631.2 | NP_001364776.1 | 1 - 2 | 181 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |