Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377631.2 | NP_001364776.1 | 4 - 5 | 799 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |