Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060456.2 | NP_492857.1 | 3 - 4 | 501 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |