Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060468.3 | NP_492869.1 | 1 - 2 | 64 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |