Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ego-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083153.5 | NP_001076622.2 | 4 - 5 | 355 | 68 | |
NM_001083154.4 | NP_001076623.1 | 3 - 4 | 310 | 68 | ||
NM_001264705.3 | NP_001251634.1 | 3 - 4 | 323 | 68 | ||
GenBank mRNA | EF378622.1 | - | 3 - 4 | 310 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |