Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrm-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060574.5 | NP_492975.3 | 3 - 4 | 701 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |