Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060630.6 | NP_493031.1 | 5 - 6 | 889 | 83 | |
GenBank mRNA | AY559747.1 | - | 4 - 5 | 739 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |