Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abt-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377649.1 | NP_001364572.1 | 18 - 19 | 4261 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |