Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060688.2 | NP_493089.2 | 9 - 10 | 1544 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |