Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047222.6 | NP_001040687.1 | 6 - 7 | 719 | 62 | |
NM_001047223.5 | NP_001040688.1 | 3 - 4 | 281 | 62 | ||
GenBank mRNA | AF070968.1 | - | 6 - 7 | 718 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |