Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026496.3 | NP_001021667.1 | 6 - 7 | 1251 | 72 | |
NM_001026497.4 | NP_001021668.1 | 5 - 6 | 950 | 72 | ||
GenBank mRNA | AY204180.1 | - | 6 - 7 | 1273 | 72 | |
AY204181.1 | - | 5 - 6 | 972 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |