Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060942.5 | NP_493343.1 | 3 - 4 | 470 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |