Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136318.5 | NP_001129790.1 | 4 - 5 | 1101 | 72 | |
NM_060942.5 | NP_493343.1 | 6 - 7 | 1406 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |