Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndx-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060971.3 | NP_493372.1 | 4 - 5 | 382 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |