Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aex-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060975.5 | NP_493376.1 | 4 - 5 | 464 | 51 | |
GenBank mRNA | AB112930.1 | - | 4 - 5 | 460 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |