Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agef-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026627.7 | NP_001021798.2 | 13 - 14 | 3782 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |