Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agef-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026627.7 | NP_001021798.2 | 1 - 2 | 166 | 71 | |
NM_001026628.5 | NP_001021799.1 | 1 - 2 | 139 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |