Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060987.5 | NP_493388.1 | 4 - 5 | 1053 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |