Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061012.6 | NP_493413.1 | 2 - 3 | 408 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |