Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zoo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265447.3 | NP_001252376.1 | 9 - 10 | 2610 | 89 | |
NM_001265448.4 | NP_001252377.1 | 8 - 9 | 2513 | 89 | ||
NM_001265449.3 | NP_001252378.1 | 5 - 6 | 2237 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |