Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lev-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026526.6 | NP_001021697.1 | 7 - 8 | 904 | 86 | |
NM_001381263.2 | NP_001367007.1 | 6 - 7 | 724 | 86 | ||
NM_001383761.1 | 9 - 10 | 1122 | 86 | |||
NM_001383762.1 | 7 - 8 | 904 | 86 | |||
NM_001383763.1 | 8 - 9 | 990 | 86 | |||
GenBank mRNA | AF298180.1 | - | 7 - 8 | 903 | 86 | |
D38541.1 | - | 9 - 10 | 1051 | 86 | ||
LC215398.1 | - | 7 - 8 | 903 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |