Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exoc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026531.5 | NP_001021702.1 | 6 - 7 | 1798 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |